Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART™ Water Analysis Laboratory

Shop All LaMotte Company Products


Click to view available options
Model:
SCL-04, Reagent Refill
SCL-04, without pH and conductivity meters
SCL-05, 120 V/60 Hz
SCL-05, Reagent Refill
Type:
Reagent Refill
Water Analysis Kit
Description
Includes
Includes digital meters to measure pH, conductivity, and TDS
- Portable.
- Measures 24 water quality parameters for pollution detection, environmental studies, and industrial water and wastes.
- Provide results directly in ppm for 6 additional factors.
- Programmed to provide direct readouts for over 40 factors using optional reagent packages.
Specifications
Specifications
pH Range | 0 to 14 |
Volume (Metric) | 500 mL |
For Use With (Application) | Pollution detection, environmental studies and industrial water and wastes |
For Use With (Equipment) | SMART colorimeter |
Includes | Digital meters to measure pH, conductivity, and TDS |
Type | Reagent Refill |
Model | SCL-05, Reagent Refill |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction