Learn More
LaMotte™ SMART™ Water Analysis Laboratory
Click to view available options
Model:
SCL-04, Reagent Refill
SCL-04, without pH and conductivity meters
SCL-05, 120 V/60 Hz
SCL-05, Reagent Refill
Type:
Reagent Refill
Water Analysis Kit
Description
Includes
Includes digital meters to measure pH, conductivity, and TDS
- Portable.
- Measures 24 water quality parameters for pollution detection, environmental studies, and industrial water and wastes.
- Provide results directly in ppm for 6 additional factors.
- Programmed to provide direct readouts for over 40 factors using optional reagent packages.
Specifications
Specifications
For Use With (Application) | Pollution detection, environmental studies and industrial water and wastes |
For Use With (Equipment) | SMART colorimeter |
Includes | Digital meters to measure pH, conductivity, and TDS |
Type | Reagent Refill |
Model | SCL-04, Reagent Refill |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov