Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART™ Water Analysis Laboratory
SDPSCI_ED

Catalog No. p-4018425
Click to view available options
Model:
SCL-04, Reagent Refill
SCL-04, without pH and conductivity meters
SCL-05, 120 V/60 Hz
SCL-05, Reagent Refill
Type:
Reagent Refill
Water Analysis Kit

Includes

Includes digital meters to measure pH, conductivity, and TDS

The SMART™ Water Analysis Laboratory features a digital colorimeter that analyzes test sample color reactions and provides direct readouts for 15 factors.

  • Portable.
  • Measures 24 water quality parameters for pollution detection, environmental studies, and industrial water and wastes.
  • Provide results directly in ppm for 6 additional factors.
  • Programmed to provide direct readouts for over 40 factors using optional reagent packages.

Specifications

Volume (Metric) 250 mL
For Use With (Application) Pollution detection, environmental studies and industrial water and wastes
For Use With (Equipment) SMART colorimeter
Includes Digital meters to measure pH, conductivity, and TDS
Type Water Analysis Kit
Model SCL-04, without pH and conductivity meters
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.