Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMP-1/CD107a Antibody (LY1C6), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1001952AF405
Description
LAMP-1/CD107a Monoclonal specifically detects LAMP-1/CD107a in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
LAMP-1/CD107a | |
Monoclonal | |
Alexa Fluor 405 | |
CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects approx 120kDa. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
LY1C6 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
LAMP1 | |
Rat liver lysosomal membrane preparations | |
0.1 mL | |
Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites | |
3916 | |
Human, Mouse, Rat, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction