Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMP-2/CD107b Antibody (H4B4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222217AF647
Description
LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human, Mouse, Chicken, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.Specifications
LAMP-2/CD107b | |
Monoclonal | |
Alexa Fluor 647 | |
P13473 | |
LAMP2 | |
LAMP-2/CD107b Antibody (H4B4) was made using a human adherent spleen cells. | |
Protein G purified | |
RUO | |
Primary | |
Human. Does not react with Mouse or Rat. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
H4B4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated | |
CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein | |
Mouse | |
45 kDa | |
0.1 mL | |
Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones | |
3920 | |
Human, Mouse, Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction