Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-2/CD107b Antibody (H4B4) - BSA Free, R&D Systems™
SDP

Catalog No. p-4966337 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222217SS 0.025 mL
NBP222217 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222217SS Supplier R&D Systems Supplier No. NBP222217SS

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody has been used in 23 publications

LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Chicken, Human, Mouse, Rat (Negative) samples. It is validated for CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, IF/IHC, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen LAMP-2/CD107b
Applications CyTOF, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone H4B4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:25, Flow Cytometry 1 ug per million cells, ELISA, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, Immunoblotting reported in scientific literature (PMID 29643474), CyTOF-ready, Knockout Validated
Gene Accession No. P13473
Gene Alias CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein
Gene Symbols LAMP2
Host Species Mouse
Immunogen LAMP-2/CD107b Antibody (H4B4) was made using a human adherent spleen cells.
Molecular Weight of Antigen 45 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3920
Test Specificity Human. Does not react with Mouse or Rat.
Target Species Chicken, Human, Mouse, Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.