Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP1 Mouse anti-Human, Clone: 5H6, Abnova™

Catalog No. 89934455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-455 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-455 Supplier Abnova Corporation Supplier No. MAB12501
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human LAMP1.

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may also play a role in tumor cell metastasis. [provided by RefSeq]

Specifications

Antigen LAMP1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 5H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human LAMP1.
Dilution Immunofluorescence (1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS (10mM sodium azide)
Gene Alias CD107a/LAMPA/LGP120
Gene Symbols LAMP1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human LAMP1.
Purification Method Affinity purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3916
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.