Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lamp1, Mouse, Unlabeled, Clone: LY1C6,

Catalog No. 428017100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
42-801-7100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-801-7100UG Supplier MilliporeSigma™ Supplier No. 428017100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamp1
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LY1C6
Conjugate Unconjugated
Formulation In PBS containing 50% glycerol, pH 7.2, ≤0.09% Sodium Azide.
Host Species Mouse
Immunogen rat liver lysosomal membranes
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage -20°C
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.