Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LAMP1 |
---|---|
Concentration | 0.48 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-100-2822
![]() ![]() |
Proteintech Group Inc
219971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Lamp1, also known as lysosome associated membrane protein 1, is a heavily glycosylated membrane protein of the lysosome. It contains a putative signal peptide, 18 sites for N-linked glycosylation, a single membrane-spanning segment and a short (11 amino acid) cytosolic tail.Specifications
LAMP1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P11279 | |
3916 | |
LAMP1 Fusion Protein Ag17367 | |
Primary | |
-20°C | |
LAMP1 |
0.48 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD107a, LAMP 1, LAMP1, LAMPA, LGP120 | |
LAMP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title