Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP2 Monoclonal Antibody (GL2A7), QED Bioscience™
SDP

Catalog No. 502278173
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8173 Supplier Leinco Technologies Inc Supplier No. 56272100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LAMP2 Monoclonal antibody specifically detects LAMP2 in Mouse, Rabbit samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen LAMP2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone GL2A7
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene LAMP2
Gene Accession No. P17047
Gene Alias CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2
Host Species Rat
Immunogen Gluteraldehyde-fixed mouse liver lysosomes
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100350946, 16784
Target Species Mouse, Rabbit
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.