Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP2 Mouse anti-Human, Mouse, Clone: 157CT16.6.6, Abnova™

Catalog No. 89934456 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-456 Supplier Abnova Corporation Supplier No. MAB12304
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human LAMP2.

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome. Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins. [provided by RefSeq]

Specifications

Antigen LAMP2
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 157CT16.6.6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human LAMP2.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:16000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene Alias CD107b/LAMPB/LGP110
Gene Symbols LAMP2
Host Species Mouse
Immunogen Recombinant protein corresponding to human LAMP2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.