Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547576
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547576 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547576 Supplier Invitrogen™ Supplier No. PA547576
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human LAMP is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMP3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene LAMP3
Gene Accession No. Q9UQV4
Gene Alias 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403
Gene Symbols LAMP3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DC-LAMP Lumenal Domain Asp21-Thr381.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.