Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728129 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8129 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8129 Supplier Proteintech Group Inc Supplier No. 126321AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysosome-associated membrane glycoprotein 3 (LAMP3, Lamp3) is a protein that in humans is encoded by theLAMP3 gene. This gene is mapped to chromosome 3q26.3-q27. It is one of the lysosome-associated membrane glycoproteins. Dendritic cells (DCs) are the most potent antigen-presenting cells.

Specifications

Antigen LAMP3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LAMP3
Gene Accession No. Q5XI99, Q7TST5, Q9UQV4
Gene Alias CD208, DC LAMP, DCLAMP, LAMP, LAMP 3, LAMP3, Protein TSC403, TSC403
Gene Symbols LAMP3
Host Species Rabbit
Immunogen LAMP3 Fusion Protein Ag3325
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239739, 27074, 303801
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.