Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMP3 Recombinant Rabbit Monoclonal Antibody (020)
GREENER_CHOICE

Catalog No. PIMA531018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531018 Supplier Invitrogen™ Supplier No. MA531018
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CD208/DC-LAMP.

Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMP3
Applications ELISA
Classification Recombinant Monoclonal
Clone 020
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LAMP3
Gene Accession No. Q9UQV4
Gene Alias 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403
Gene Symbols LAMP3
Host Species Rabbit
Immunogen Recombinant Human CD208/DC-LAMP Protein (Lys28-Thr381).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.