Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMR1 Mouse anti-Human, Clone: 5D4, Bioss
SDP

Catalog No. 501979874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9874 Supplier Bioss Supplier No. BSM0940M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Many of the effects of laminin are mediated through interactions with cell surface receptors. These receptors include members of the integrin family, as well as non-integrin laminin-binding proteins. This gene encodes a high-affinity, non-integrin family, laminin receptor 1. This receptor has been variously called 67 kD laminin receptor, 37 kD laminin receptor precursor (37LRP) and p40 ribosome-associated protein. The amino acid sequence of laminin receptor 1 is highly conserved through evolution, suggesting a key biological function. It has been observed that the level of the laminin receptor transcript is higher in colon carcinoma tissue and lung cancer cell line than their normal counterparts. Also, there is a correlation between the upregulation of this polypeptide in cancer cells and their invasive and metastatic phenotype. Multiple copies of this gene exist, however, most of them are pseudogenes thought to have arisen from retropositional events. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen LAMR1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5D4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RPSA
Gene Accession No. P08865
Gene Alias 37 kDa laminin receptor; 37 kDa laminin receptor precursor; 37 kDa laminin receptor precursor {ECO:0000255; 37 kDa oncofetal antigen; 37/67 kDa laminin receptor; 37/67 kDa laminin receptor {ECO:0000255; 37LRP; 37LRP {ECO:0000255; 40S ribosomal protein SA; 40S ribosomal protein SA {ECO:0000255; 67 kDa laminin receptor; 67 kDa laminin receptor {ECO:0000255; 67kDa; 67LR; 67LR {ECO:0000255; AL022858; Colon carcinoma laminin-binding protein; HAMAP-Rule:MF_03016}; ICAS; LAMBR; Laminin receptor 1; laminin receptor 1 (67kD, ribosomal protein SA); laminin receptor 1 (ribosomal protein SA); laminin receptor 1 {ECO:0000255; Laminin-binding protein precursor p40; laminin-binding protein precursor p40 {ECO:0000255; LamR; lamR {ECO:0000255; Lamr1; Lamrl1; LBP; LBP/p40; LBP/p40 {ECO:0000255; LRP; LRP/LR; LRP/LR {ECO:0000255; MLR; Multidrug resistance-associated protein MGr1-Ag; NEM/1CHD4; OFA/iLRP; P40; P40-3; P40-3, functional; P40-8; P40-8, functional; ribosomal protein SA; Rpsa; SA; Small ribosomal subunit protein uS2
Gene Symbols RPSA
Host Species Mouse
Immunogen KLH conjugated synthetic peptide derived from middle of human LAMR1.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3921
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.