Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMTOR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551792 Supplier Invitrogen™ Supplier No. PA551792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEDSDQDREE RKLLLDPSSP PTKALNGAEP NYHSLPSART DEQALLSSIL AKTASNIIDV SAADSQGMEQ HEYMDRARQY STRLAVLSSS LTHWKKLPPL PSLTSQPHQV LASEPIPFSD LQQVSRIAAY AYSALSQIRV DAKEELVV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

The Ragulator complex is comprised of three proteins, C11orf59, ROBLD3, and MAPKSP1, which targets mTORC1 to lysosomal surfaces. This normal endosomal trafficking is taken advantage of by certain viruses, including coronavirus, which has increased replication when C11orf59 is interfered with. C11orf59 is positively regulated by cellular cholesterol depletion and negatively regulated by cellular cholesterol loading. It is involved in LDL uptake and cholesterol homeostasis. C11orf59 and PPAR-alpha work together to control lipid and carbohydrate metabolism in diabetes. Nearly all effects of C11orf59 are associated with mTORC1 kinase growth promotion, and disregulation of the normal mTOR pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMTOR1
Gene Accession No. Q6IAA8
Gene Alias 2400001E08Rik; C11orf59; Lamtor1; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; late endosomal/lysosomal adaptor, MAPK and MTOR activator 1; lipid raft adaptor protein p18; p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF Rho; p27RF-Rho; Pdro; PP7157; Protein associated with DRMs and endosomes; ragulator complex protein LAMTOR1; ragulator complex protein PDRO; Ragulator1; RhoA activator C11orf59; RhoA activator C11orf59 homolog
Gene Symbols LAMTOR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAMTOR1. Recombinant protein control fragment (Product #RP-95926).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55004
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.