Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMTOR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578516 Supplier Thermo Scientific Supplier No. PA578516
Add to Cart
Add to Cart

LAMTOR1 Polyclonal Antibody

LAMTOR1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene LAMTOR1
Gene Accession No. Q6IAA8, Q9CQ22
Gene Alias 2400001E08Rik; C11orf59; Lamtor1; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; late endosomal/lysosomal adaptor, MAPK and MTOR activator 1; lipid raft adaptor protein p18; p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF Rho; p27RF-Rho; Pdro; PP7157; Protein associated with DRMs and endosomes; ragulator complex protein LAMTOR1; ragulator complex protein PDRO; Ragulator1; RhoA activator C11orf59; RhoA activator C11orf59 homolog
Gene Symbols LAMTOR1
Host Species Rabbit
Immunogen Full length human LAMTOR1 recombinant protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55004, 66508
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.