Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAMTOR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115491
Description
Antibody detects endogenous levels of total LAMTOR1.
The Ragulator complex is comprised of three proteins, C11orf59, ROBLD3, and MAPKSP1, which targets mTORC1 to lysosomal surfaces. This normal endosomal trafficking is taken advantage of by certain viruses, including coronavirus, which has increased replication when C11orf59 is interfered with. C11orf59 is positively regulated by cellular cholesterol depletion and negatively regulated by cellular cholesterol loading. It is involved in LDL uptake and cholesterol homeostasis. C11orf59 and PPAR-alpha work together to control lipid and carbohydrate metabolism in diabetes. Nearly all effects of C11orf59 are associated with mTORC1 kinase growth promotion, and disregulation of the normal mTOR pathway.
Specifications
LAMTOR1 | |
Polyclonal | |
Unconjugated | |
LAMTOR1 | |
2400001E08Rik; C11orf59; Lamtor1; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1; late endosomal/lysosomal adaptor, MAPK and MTOR activator 1; lipid raft adaptor protein p18; p18; p27kip1 releasing factor from RhoA; p27Kip1-releasing factor from RhoA; p27RF Rho; p27RF-Rho; Pdro; PP7157; Protein associated with DRMs and endosomes; ragulator complex protein LAMTOR1; ragulator complex protein PDRO; Ragulator1; RhoA activator C11orf59; RhoA activator C11orf59 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
308869, 55004, 66508 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q6IAA8, Q6P791, Q9CQ22 | |
LAMTOR1 | |
A synthesized peptide derived from human LAMTOR1(Accession Q6IAA8), corresponding to amino acid residues E10-K60. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction