Learn More
LAMTOR1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305284A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Orangutan
The Ragulator complex is comprised of three proteins, C11orf59, ROBLD3, and MAPKSP1, which targets mTORC1 to lysosomal surfaces. This normal endosomal trafficking is taken advantage of by certain viruses, including coronavirus, which has increased replication when C11orf59 is interfered with. C11orf59 is positively regulated by cellular cholesterol depletion and negatively regulated by cellular cholesterol loading. It is involved in LDL uptake and cholesterol homeostasis. C11orf59 and PPAR-alpha work together to control lipid and carbohydrate metabolism in diabetes. Nearly all effects of C11orf59 are associated with mTORC1 kinase growth promotion, and disregulation of the normal mTOR pathway.Specifications
LAMTOR1 | |
Polyclonal | |
Unconjugated | |
LAMTOR1 | |
PP7157, C11orf59, PDRO, Ragulator1, p18, p27RF-Rho, RhoA activator C11orf59, late endosomal/lysosomal adaptor and MAPK and MTOR activator 1, lipid raft adaptor protein p18, p27Kip1-releasing factor from RhoA, p27kip1 releasing factor from RhoA, protein associated with DRMs and endosomes, ragulator complex protein LAMTOR1, ragulator complex protein PDRO | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55004 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6IAA8 | |
LAMTOR1 | |
Between 111 and 161 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.