Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAMTOR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA538033
Description
A recommended positive control is HepG2 cell lysate. At least two isoforms of LAMTOR2 are known to exist; this antibody will detect both isoforms. LAMTOR2 antibody is not expected to cross-react with other LAMTOR family proteins.
The late endosomal/lysosomal adaptor MAPK and MTOR activator 2 (LAMTOR2) protein belongs to the LAMTOR family of proteins, and together with LAMTOR3 and the MAPK1 and ERK kinase 1 (MEK1) localizes to late endosomes where it is required for the efficient activation of ERK signaling. This complex is involved in the regulation of late endosomal traffic and cellular proliferation and plays a role in cellular host defense against Salmonella infection.
Specifications
LAMTOR2 | |
Polyclonal | |
Unconjugated | |
LAMTOR2 | |
2010111E04Rik; AL022628; ENDAP; Endosomal adaptor protein p14; HSPC003; LAMTOR2; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 2; late endosomal/lysosomal adaptor, MAPK and MTOR activator 2; late endosomal/lysosomal MP1 interacting protein; late endosomal/lysosomal Mp1-interacting protein; MAPBP-interacting protein; Mapbpip; MAPKSP1 adaptor protein; MAPKSP1AP; mitogen activated protein binding protein interacting protein; mitogen-activated protein binding protein interacting protein; mitogen-activated protein-binding protein-interacting protein; mitogen-activated protein-binding protein-interacting protein (MAPBPIP); P14; Rab25; ragulator complex protein LAMTOR2; Ragulator2; RGD1562501; roadblock domain containing 3; roadblock domain-containing protein 3; ROBLD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28956, 295234, 83409 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9JHS3, Q9Y2Q5 | |
LAMTOR2 | |
18 amino acid peptide near the center of human LAMTOR2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction