Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMTOR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552056 Supplier Invitrogen™ Supplier No. PA552056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPKALTQVLS QANTGGVQST LLLNNEGSLL AYSGYGDTDA RVTAAIASNI WAAYDRNGNQ AFNEDNLKFI LMDCMEGRVA ITRVANLLLC MYAKETVGFG MLKAKAQALV QYLEEP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The late endosomal/lysosomal adaptor MAPK and MTOR activator 2 (LAMTOR2) protein belongs to the LAMTOR family of proteins, and together with LAMTOR3 and the MAPK1 and ERK kinase 1 (MEK1) localizes to late endosomes where it is required for the efficient activation of ERK signaling. This complex is involved in the regulation of late endosomal traffic and cellular proliferation and plays a role in cellular host defense against Salmonella infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMTOR2
Gene Accession No. Q9Y2Q5
Gene Alias 2010111E04Rik; AL022628; ENDAP; Endosomal adaptor protein p14; HSPC003; LAMTOR2; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 2; late endosomal/lysosomal adaptor, MAPK and MTOR activator 2; late endosomal/lysosomal MP1 interacting protein; late endosomal/lysosomal Mp1-interacting protein; MAPBP-interacting protein; Mapbpip; MAPKSP1 adaptor protein; MAPKSP1AP; mitogen activated protein binding protein interacting protein; mitogen-activated protein binding protein interacting protein; mitogen-activated protein-binding protein-interacting protein; mitogen-activated protein-binding protein-interacting protein (MAPBPIP); P14; Rab25; ragulator complex protein LAMTOR2; Ragulator2; RGD1562501; roadblock domain containing 3; roadblock domain-containing protein 3; ROBLD3
Gene Symbols LAMTOR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LAMTOR2. Recombinant protein control fragment (Product #RP-102170).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28956
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.