Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMTOR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114758 Supplier Invitrogen™ Supplier No. PA5114758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LAMTOR3.

This gene encodes a scaffold protein that functions in the extracellular signal-regulated kinase (ERK) cascade. The protein is localized to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. Studies of the orthologous gene in mouse indicate that it regulates late endosomal traffic and cell proliferation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene LAMTOR3
Gene Accession No. O88653, Q5U204, Q9UHA4
Gene Alias AW556229; hypothetical protein LOC553597; lamtor3; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 3; late endosomal/lysosomal adaptor, MAPK and MTOR activator 3; Map2k1ip1; MAPBP; MAPK scaffold protein 1; MAPKSP1; MEK binding partner 1; MEK partner 1; MEK-binding partner 1; mitogen activated protein, binding protein; mitogen-activated protein kinase kinase 1 interacting protein 1; mitogen-activated protein kinase kinase 1-interacting protein 1; Mitogen-activated protein kinase scaffold protein 1; mitogen-activated protein kinase scaffold protein 1; ragulator complex protein LAMTOR3; Mp1; PRO0633; PRO2783; Ragulator complex protein LAMTOR3; Ragulator3; zgc:110207
Gene Symbols LAMTOR3
Host Species Rabbit
Immunogen A synthesized peptide derived from human LAMTOR3(Accession Q9UHA4), corresponding to amino acid residues A36-L86.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362045, 56692, 8649
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.