Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAMTOR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578680
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578680 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578680 Supplier Invitrogen™ Supplier No. PA578680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HeLa, SKNSH Predicted Reactivity: Mouse (95%), Rat (96%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

LAMTOR4 is a part of the Ragulator complex it is involved in amino acid sensing and activation of mTORC1, a signaling complex promoting cell growth in response to growth factors, energy levels, and amino acids. LAMTOR4 is activated by amino acids through a mechanism involving the lysosomal V-ATPase, the Ragulator functions as a guanine nucleotide exchange factor activating the small GTPases Rag. LAMTOR4 is activated Ragulator and Rag GTPases function as a scaffold recruiting mTORC1 to lysosomes where it is in turn activated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMTOR4
Applications Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene LAMTOR4
Gene Accession No. Q0VGL1
Gene Alias 0910001L09Rik; AV006840; C7orf59; Lamtor4; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 4; late endosomal/lysosomal adaptor, MAPK and MTOR activator 4; ragulator complex protein LAMTOR4; Ragulator complex protein LAMTOR4, N-terminally processed; RGD1309735; UPF0539 protein C7orf59; UPF0539 protein C7orf59 homolog
Gene Symbols LAMTOR4
Host Species Rabbit
Immunogen Full length human LAMTOR4 recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.