Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAMTOR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554301
Description
Immunogen sequence: MTSALTQGLE RIPDQLGYLV LSEGAVLASS GDLENDEQAA SAISELVSTA CGFRLHRGMN VPFKRLSVVF GEHTLLVTVS GQRVFVVKRQ NRGREPIDV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
LAMTOR4 is a part of the Ragulator complex it is involved in amino acid sensing and activation of mTORC1, a signaling complex promoting cell growth in response to growth factors, energy levels, and amino acids. LAMTOR4 is activated by amino acids through a mechanism involving the lysosomal V-ATPase, the Ragulator functions as a guanine nucleotide exchange factor activating the small GTPases Rag. LAMTOR4 is activated Ragulator and Rag GTPases function as a scaffold recruiting mTORC1 to lysosomes where it is in turn activated.
Specifications
LAMTOR4 | |
Polyclonal | |
Unconjugated | |
LAMTOR4 | |
0910001L09Rik; AV006840; C7orf59; Lamtor4; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 4; late endosomal/lysosomal adaptor, MAPK and MTOR activator 4; ragulator complex protein LAMTOR4; Ragulator complex protein LAMTOR4, N-terminally processed; RGD1309735; UPF0539 protein C7orf59; UPF0539 protein C7orf59 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389541 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VGL1 | |
LAMTOR4 | |
Recombinant protein corresponding to Human LAMTOR4. Recombinant protein control fragment (Product #RP-92832). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction