Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LANCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557107 Supplier Invitrogen™ Supplier No. PA557107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQRAFPNPY ADYNKSLAEG YFDAAGRLTP EFSQRLTNKI RELLQQM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LANCL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LANCL1
Gene Accession No. O43813, O89112, Q9QX69
Gene Alias 40 kDa erythrocyte membrane protein; AW124738; G protein-coupled receptor 69; G protein-coupled receptor 69A; Glutathione S-transferase LANCL1; Gpr69; Gpr69a; LanC (bacterial lantibiotic synthetase component C)-like 1; LanC (bacterial lantibiotic synthetase component); LanC lantibiotic synthetase component C-like 1; LanC lantibiotic synthetase component C-like 1 (bacterial); LanC like 1; LANCL1; LanC-like 1; LanC-like 1(bacterial); lanC-like protein 1; p40
Gene Symbols LANCL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LANCL1. Recombinant protein control fragment (Product #RP-96014).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10314, 114515, 14768
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.