Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LANCL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $665.78
Specifications
| Antigen | LANCL2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
LANCL2 Polyclonal specifically detects LANCL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LANCL2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| G protein-coupled receptor 69B, GPR69B, LanC (bacterial lantibiotic synthetase component C)-like 2, LanC lantibiotic synthetase component C-like 2 (bacterial), lanC-like protein 2, TASPMGC87139, Testis-specific adriamycin sensitivity protein | |
| LANCL2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 55915 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ALLYLNTEIGPGTVCESAIKEVVNAIIESGKTLSREERKTERCPLLYQWHRKQYVGAAHGMAGIYYMLMQPAAKVDQETLTEMVKPSIDY | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title