Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANCL2 Mouse anti-Human, Clone: 392, MilliporeSigma™

Catalog No. MABS1290MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1290MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1290MI Supplier MilliporeSigma Supplier No. MABS1290

Mouse Monoclonal Antibody

Detect LANCL2 using this mouse monoclonal Anti-LANCL2 Antibody, clone 392. It is used in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LANCL2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 392
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LANCL2,GPR69B,TASP
Host Species Mouse
Immunogen GST-tagged full length human recombinant LANCL2 protein.
Purification Method Protein A Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_061167
Test Specificity Clone 392 detected LANCL2 overexpressed in HEK293 cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.