Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Langerin associated lectin Antibody (109B8.02), Alexa Fluor™ 546, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0350A546100
Description
Langerin associated lectin Monoclonal specifically detects Langerin associated lectin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Langerin associated lectin | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Human | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
109B8.02 | |
Alexa Fluor 546 | |
LAL, Langerin-associated-lectin | |
Langerin-associated lectin/Human Kupffer cell receptor homolog was used as immunogen. | |
0.1 mg | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction