Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Langerin/CD207 Antibody (310F7.02), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 15 publications
Supplier: Novus Biologicals DDX0361P100
Description
Langerin/CD207 Monoclonal specifically detects Langerin/CD207 in Human, Mouse, Rat, Porcine, Chinese Hamster samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).Specifications
Langerin/CD207 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Cell Surface), Flow (Intracellular) | |
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Chinese Hamster | |
IgG1 |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
310F7.02 | |
Unconjugated | |
NP_001207 | |
CD207 | |
Langerin/CD207 was used as immunogen. | |
0.1 mg | |
metabolism, Signal Transduction | |
50489 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction