Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Langerin/CD207 Antibody (918A11), Alexa Fluor™ 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals DDX0376A488
Description
Langerin/CD207 Monoclonal specifically detects Langerin/CD207 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).Specifications
Langerin/CD207 | |
Monoclonal | |
Alexa Fluor 488 | |
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin | |
Rat | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
918A11 | |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Flow (Cell Surface) 1:10-1:1000 | |
CD207 | |
Langerin/CD207 was used as immunogen. | |
0.1 mg | |
metabolism, Signal Transduction | |
50489 | |
Store at -20C in the dark. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction