Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Mouse anti-Human, Alexa Fluor 700, Clone: 343828, R&D Systems™

Catalog No. IC2088N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2088N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2088N100 Supplier R&D Systems Supplier No. IC2088N100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Langerin/CD207
Applications Flow Cytometry
Classification Monoclonal
Clone 343828
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UJ71
Gene Alias CD207, CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Gene Symbols CD207
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Langerin/CD207 Tyr64-Pro328 Accession # Q9UJ71
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50489
Test Specificity Detects human Langerin/CD207 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.