Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lano Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176678PCP
Description
Lano Polyclonal antibody specifically detects Lano in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Lano | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
dJ523E19.1, FLJ10775, FLJ11834, LANOLANO adapter protein, LAP (leucine-rich repeats and PDZ) and no PDZ protein, LAP and no PDZ protein, leucine rich repeat containing 1, leucine-rich repeat-containing 1, leucine-rich repeat-containing protein 1 | |
Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human Lano. The immunogen is located within the last 50 amino acids of Lano. Amino Acid Squence: KNEVNHAIDRVTTSV | |
0.1 mL | |
Cytokines and Growth Factors, Immunology, Neuroscience | |
55227 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction