Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanolin (USP), Fisher Chemical

Catalog No. l7500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Amber Glass

Wool Fat

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-54-0, 7732-18-5
MDL Number MFCD00081740
Synonym dmt-2'o-tbdms-rc ac phosphoramidite, dmt-2'o-tbdms-rc ac phosphoramidite, configured for abi, dmt-2'o-tbdms-rc ac phosphoramidite, configured for mermade, ac-c-ce phosphoramidite, dmt-2'o-tbdms-rc ac amidite, dmt-2'o-tbdms-rc ac pharmadite r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for ??kta r and oligopilot r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for perkinelmer, configured for polygen, dmt-2'o-tbdms-rc ac phosphoramidite, configured for polygen, configured for perkinelmer

Specifications

Alkalinity Pass Test
Acidity Pass Test
Melting Point 36.1°C
Color Yellow
Ignition Residue 0.1% max.
Packaging Amber Glass
Quantity 500 g
Grade USP
Physical Form Solid
Chemical Name or Material Lanolin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.