Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Lanolin, USP, Spectrum™ Chemical
 
    
    
    
        
            Shop All Spectrum Chemical Manufacturing Corporation Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                430 g
                            
                        
                            
                                6 x 430 g
                            
                        
                            
                                18 kg
                            
                        
                            
                                180 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Poly Bottle
                            
                        
                            
                                Poly Drum
                            
                        
                            
                                Poly Pail
                            
                        
                    Description
Spectrum™ Chemical Lanolin, USP  is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
                
    
            Specifications
Specifications
| CAS Max % | 100% | 
| Ignition Residue | 0.1% | 
| Melting Range | 38°-44°C | 
| Packaging | Poly Bottle | 
| Quantity | 430 g | 
| Grade | USP | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                