Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lanolin, USP, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
430 g
6 x 430 g
18 kg
180 kg
Packaging:
Poly Bottle
Poly Drum
Poly Pail
Description
Spectrum™ Chemical Lanolin, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Melting Range | 38°-44°C |
| Packaging | Poly Bottle |
| Quantity | 6 x 430 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction