Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum chloride, Releasing Agent Solution, Specpure™

Catalog No. aa39036ap
Click to view available options
Quantity:
500 mL

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

Lanthanum chloride is a precursor for synthesis of lanthanum phosphate nano rods and used in gamma detectors. It is also used as a catalyst for high pressure oxidative chlorination of methane to chloromethane with hydrochloric acid and oxygen. In organic synthesis, lanthanum trichloride acts as lewis acid for the conversion of aldehydes to acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Physical Form Liquid
Quantity 500 mL
Solubility Information Miscible with water.
Chemical Name or Material Lanthanum chloride
Hazard Category H290-H315-H319
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.