Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) acetate sesquihydrate, REacton™, 99.99% (REO)

Catalog No. p-7048695
Click to view available options
Quantity:
50 g
250 g

CAS: 25721-92-0 | C12H24La2O15 | 686.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25721-92-0
Molecular Formula C12H24La2O15
Molecular Weight (g/mol) 686.12
MDL Number MFCD00013037
InChI Key RBPQRWPSWRTKEC-UHFFFAOYSA-H
IUPAC Name dilanthanum(3+) hexaacetate trihydrate
SMILES O.O.O.[La+3].[La+3].CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point <200°C (in -1.5H2O)
Physical Form Crystalline
Quantity 50 g
Linear Formula La(OOCCH3)3·1.5H2O
Formula Weight 343.05 (316.03 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) acetate sesquihydrate
EINECSNumber 213-034-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.