Lanthanum(III) chloride heptahydrate, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer: Alfa Aesar 1126818

Quantity 50g

 View more versions of this product

Catalog No. AA1126818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-84-0
    Physical Form Wet Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula LaCl3·7H2O
    Formula Weight 371.37; 245.27 (anhydrous)
    MDL Number MFCD00149756
    Chemical Name or Material Lanthanum(III) chloride heptahydrate

    Specifications

    Formula Weight 371.37; 245.27 (anhydrous)
    Quantity 50g
    Color White
    EINECS Number 233-237-5
    Physical Form Wet Crystalline Aggregates
    Odor Odorless
    Assay Percent Range 99.99% (REO)
    Molecular Formula LaCl3·7H2O
    Appearance White wet crystalline aggregates
    MDL Number MFCD00149756
    CAS 10025-84-0
    Health Hazard 1 H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
    Chemical Name or Material Lanthanum(III) chloride heptahydrate

    H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit