Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride heptahydrate, ACS

Catalog No. p-7049106
Click to view available options
Quantity:
50 g
250 g

Catalog No. AA1291618

Supplier: Thermo Scientific Chemicals 01291618

Only null left
Add to Cart

CAS: 10025-84-0 | Cl3H14LaO7 | 371.36 g/mol

Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91°C (decomposition)
Physical Form Crystalline
Quantity 50 g
Merck Index 14,5363
Solubility Information Soluble in ethanol and water.
Formula Weight 371.37 (245.27 Anhydrous)
Grade ACS
Chemical Name or Material Lanthanum(III) chloride heptahydrate, For analysis ACS
Hazard Category H290-H317-H318
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
EINECSNumber 233-237-5
RTECSNumber OE4470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.