Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. AA1366318
Click to view available options
Quantity:
50 g

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]

Specifications

Melting Point 1493°C
Physical Form Lump
Quantity 50 g
Linear Formula LaF3
Solubility Information Insoluble in water. Soluble in strong mineral acids.
Formula Weight 195.9
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 5.936 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-252-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.