Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Alanine methyl ester hydrochloride, 98+%

Catalog No. AC393310050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle

Catalog No. AC393310050

Supplier: Thermo Scientific Chemicals 393310050

Only null left
Add to Cart

CAS: 13515-97-4 | C4H10ClNO2 | 139.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13515-97-4
Molecular Formula C4H10ClNO2
Molecular Weight (g/mol) 139.58
MDL Number MFCD00035523
InChI Key IYUKFAFDFHZKPI-UHFFFAOYNA-N
Synonym methyl 2-aminopropanoate hydrochloride, dl-alanine methyl ester hydrochloride, h-dl-ala-ome.hcl, h-dl-ala-ome hydrochloride, dl-alanine methyl ester hcl, methyl l-alaninate hcl, methyl dl-alaninate hcl, methyl dl-alaninate hydrochloride, h-dl-ala-ome hcl, methyl dl-2-aminopropanoate hydrochloride
PubChem CID 2756298
SMILES Cl.COC(=O)C(C)N

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula CH3CH(NH2)COOCH3·HCl
IUPAC Name methyl 2-aminopropanoate hydrochloride
Quantity 5 g
Formula Weight 139.58
Percent Purity ≥98.0%
Physical Form Powder
Chemical Name or Material DL-Alanine methyl ester hydrochloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.