Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO)

Catalog No. AA1126718
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg

CAS: 10277-43-7 | H12LaN3O15 | 433.01 g/mol

Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 40°C
Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water,alcohol and acids.
IUPAC Name lanthanum(3+) hexahydrate trinitrate
Formula Weight 433.01 (324.92 Anhydrous)
Odor Odorless
Appearance White crystalline
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 233-238-0
RTECSNumber OE5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.