Lanthanum(III) nitrate hexahydrate, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer: Alfa Aesar 1126706

Quantity 5g

 View more versions of this product

Catalog No. AA1126706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10277-43-7
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula La(NO3)3·6H2O
    Formula Weight 433.01 (324.92 Anhydrous)
    MDL Number MFCD00149751
    Chemical Name or Material Lanthanum(III) nitrate hexahydrate

    Specifications

    Physical Form Crystalline Aggregates
    Formula Weight 433.01 (324.92 Anhydrous)
    Quantity 5g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 233-238-0
    Odor Odorless
    Assay Percent Range 99.99% (REO)
    Molecular Formula La(NO3)3·6H2O
    Appearance White crystalline
    MDL Number MFCD00149751
    CAS 10277-43-7
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Lanthanum(III) nitrate hexahydrate

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit