Learn More
Lanthanum(III) nitrate hexahydrate, REacton™, 99.999% (REO)
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 40°C |
Color | White |
Physical Form | Crystalline |
Quantity | 500 g |
Assay Percent Range | 99.999% (REO) |
Solubility Information | Soluble in water,alcohol and acids. |
IUPAC Name | lanthanum(3+) hexahydrate trinitrate |
Formula Weight | 433.01 (324.92 Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1; Packaging Group: II |
EINECSNumber | 233-238-0 |
RTECSNumber | OE5075000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.