Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog No. AA1319618
Click to view available options
Quantity:
50 g
250 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537-03-1
Molecular Formula C6La2O12
Molecular Weight (g/mol) 541.865
MDL Number MFCD00150540
InChI Key OXHNIMPTBAKYRS-UHFFFAOYSA-H
Synonym lanthanum oxalate, lanthanum iii oxalate, acmc-1bafu, lanthanum 3+ ; oxalate, dilanthanum 3+ trioxalate ion, dilanthanum 3+ trioxalate, lanthanum iii oxalate decahydrate, reacton, ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9, lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di
PubChem CID 164654
IUPAC Name lanthanum(3+);oxalate
SMILES C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula La2(C2O4)3·10H2O
UN Number UN3288
Formula Weight 722.02 (541.87 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) oxalate decahydrate
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-656-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.