Learn More
Lanthanum(III) oxide, REacton™, 99.9% (REO)
Description
Lanthanum(III) oxide is used as a precursor to LAMOX fast ion conductors and superconductors. It is also used in the manufacture of optical glasses, piezoelectric and thermoelectric material. It is also useful for X-ray imaging intensifying screens, phosphors as well as dielectric and conductive ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 4200°C |
Melting Point | 2307°C |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.9% (REO) |
Molecular Formula | La2O3 |
Solubility Information | Insoluble in water but soluble in dilute acid. |
Formula Weight | 325.82 |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
EINECSNumber | 215-200-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.