Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) oxide, 99.9%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
1312-81-8
La2O3
325.82
MFCD00011071
1312-81-8
La2O3
325.82
MFCD00011071
for ultra trace analysis
Assay Percent Range 99.85% min. (La2O3) TREO base
Linear Formula La2O3
Merck Index 15, 5414
Solubility Information Solubility in water: insoluble
Formula Weight 325.82
Physical Form Crystals or Crystalline Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Fe2O3 10ppm max.
Loss on Ignition 3.0 % max. (1000°C)
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Boiling Point 4200.0°C
Color Blue
Melting Point 2315.0°C
Quantity 250g
Chemical Name or Material Lanthanum(III) oxide, 99.9%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 215-200-5

RTECSNumber : OE5330000

TSCA : TSCA