Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) phosphate hydrate

Catalog No. p-7058727
Click to view available options
Quantity:
25 g
100 g

CAS: 14913-14-5 | LaO4P | 233.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14913-14-5
Molecular Formula LaO4P
Molecular Weight (g/mol) 233.88
MDL Number MFCD00150248
InChI Key LQFNMFDUAPEJRY-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) phosphate
SMILES [La+3].[O-]P([O-])([O-])=O

Specifications

Physical Form Lump
Quantity 25 g
Formula Weight 233.88 (Anhydrous)
Odor Odorless
Chemical Name or Material Lanthanum (III) phosphate hydrate
EINECSNumber 237-419-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.