Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)

Catalog No. AA40292DH
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm

Form: Approx.0.20g/mm, Cast rod, diameter is approximate | CAS: 7439-91-0 | La | 138.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-91-0
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]

Specifications

Color Silver-White
Appearance Silvery-white rod
Density 6.17g/cm3 at 20°C
Quantity 25 mm
Packaging Mylar under Argon
Form Cast Rod
Assay Percent Range 99.9% (REO)
Chemical Name or Material Lanthanum, Rare Earth Oxides Content
Hazard Category H228
Hazard Statement Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-099-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.