Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-Biotin Antibody
LanthaScreen™ Eu-anti-Biotin Antibody
$1280.00 - $14280.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV5901
Encompass_Preferred
View Documents
Thermo Scientific™
PV5901
25 μg
Each for $1,280.00
Only null left
Add to Cart
PV5900
Encompass_Preferred
View Documents
Thermo Scientific™
PV5900
1 mg
Each for $14,280.00
Only null left
Add to Cart
Description
Description
A general reagent for use in assay development, the Europium-anti-biotin antibody binds to biotinylated peptide or physiological protein substrates.
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.